Learn More
Palladium Nickel gauze, 50 mesh woven from 0.1578mm (0.0062in) dia wire, Thermo Scientific Chemicals
Filtration
503.51 € - 1643.18 €
Specifications
Chemical Name or Material | Palladium Nickel gauze |
---|---|
Name Note | 50 mesh woven from 0.1578mm (0.0062 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Pd:Ni; 95:5 wt% |
MDL Number | MFCD05863544 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439920
|
Thermo Scientific Alfa Aesar
044698.FL |
75 x 75 mm |
503.51 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449920
|
Thermo Scientific Alfa Aesar
044698.GT |
150 x 150 mm |
1643.18 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Palladium Nickel gauze is used in filtrations. It is also used as a capture gauze for platinum recovery in ammonia oxidation apparatus with a pressure of 4 atm (405 kPa) and a temperature of 8500C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium Nickel gauze | |
Pd:Ni; 95:5 wt% | |
Insoluble in water. | |
Wire Cloth |
50 mesh woven from 0.1578mm (0.0062 in.) dia. wire | |
MFCD05863544 | |
50 Mesh |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H373
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only