missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium powder, APS 0.35-0.8 micron, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
0.1 g
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
UN Number | UN3089 |
Quantity | 1 g |
Percent Purity | 99.95% |
Odor | Odorless |
Particle Size | APS 0.35 to 0.8 micron |
Form | Powder |
Assay | (metals basis) |
Chemical Name or Material | Palladium |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W08J040 | Certificate of Analysis | 21/09/2023 | 047249.06,047249.77,047249.03 |
T17J025 | Certificate of Analysis | 21/07/2023 | 047249.06,047249.77,047249.03 |
U04I026 | Certificate of Analysis | 19/08/2022 | |
Z16H026 | Certificate of Analysis | 12/04/2022 | |
N03H016 | Certificate of Analysis | 08/12/2021 |
Safety and Handling


Product Identifier
- Palladium powder
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H228-Flammable solid.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Palladium, powder
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Palladium powder, APS 0.35-0.8 micron, 99.95% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction