Learn More
Palladium Silver foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding Pt), Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 042682.FF
 View more versions of this product
Product Code. 11385417
  • 117.59 € / 25mm
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Palladium Silver foil
Pd:Ag; 75:25 wt%
MFCD02091738
99.9%
≈0.36g/25x25mm
0.05mm (0.002 in.) thick
(metals basis excluding Pt)
25 x 25 mm
Odorless
Foil