missing translation for 'onlineSavingsMsg'
Learn More

Palladium standard solution, for AAS, 1 mg/ml Pd in 10% HCl, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196175000


 View more versions of this product

Product Code. 10685642

  • 162.81 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Palladium standard solution
1 mg/ml Pd in 10% HCl
Plastic bottle
Brown/Red
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
[Pd]
106.42
CHEBI:33363
1.0500g/mL
107.0°C
Soluble in water
997 to 1003 μg/mL of Pd (2+) in 10% HCl (20°C)
7440-05-3, 7647-01-0, 7732-18-5
500 mL
Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
AAS
-25.0°C
1.05
Solution
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Palladium atomic absorption standard solution

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only