missing translation for 'onlineSavingsMsg'
Learn More

Palladium wire, 0.5mm (0.02in) dia, Premion™, 99.99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11311528
Click to view available options
Quantity:
50 cm
250 cm
1000 cm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Quantity 250 cm
Percent Purity ≥99.99%
Grade Premion™
Odor Odorless
Weight ≈2.36g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Palladium wire, 0.5mm (0.02 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'