missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10731161
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-67-4
MDL Number MFCD00000742
InChI Key ARBOVOVUTSQWSS-UHFFFAOYSA-N
Synonym palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
PubChem CID 8206
IUPAC Name hexadecanoyl chloride
SMILES CCCCCCCCCCCCCCCC(=O)Cl

Specifications

Acidity 1% max. (as palmitic acid)
Density 0.9000g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C16H31ClO
Linear Formula CH3(CH2)14COCl
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Quantity 5 mL
Beilstein 02, 374
Molecular Weight (g/mol) 274.873
Formula Weight 274.88
Percent Purity 98 to 101% (Argentometry), ≥97.5%(GC)
Physical Form Liquid
Chemical Name or Material Palmitoyl chloride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'