Learn More
Pamoic acid, 99%, Thermo Scientific Chemicals
34.97 € - 113.57 €
Chemical Identifiers
CAS | 130-85-8 |
---|---|
Molecular Formula | C23H16O6 |
Molecular Weight (g/mol) | 388.38 |
MDL Number | MFCD00004079 |
InChI Key | WLJNZVDCPSBLRP-UHFFFAOYSA-N |
Synonym | pamoic acid, embonic acid, 4,4'-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7rrq8qz38n, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 7rrq8qz38n, pamoic acid 98+% hplc, 4,4'-methylen-bis-3-hydroxy-2-naphthoesaeure |
PubChem CID | 8546 |
ChEBI | CHEBI:50186 |
IUPAC Name | 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid |
SMILES | OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1 |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
11492218
|
Thermo Scientific Alfa Aesar
A15207.18 |
50 g |
34.97 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402228
|
Thermo Scientific Alfa Aesar
A15207.30 |
250 g |
113.57 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
Solubility
Soluble in nitrobenzene, pyridine
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Identificatori chimici
130-85-8 | |
388.38 | |
WLJNZVDCPSBLRP-UHFFFAOYSA-N | |
8546 | |
4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid |
C23H16O6 | |
MFCD00004079 | |
pamoic acid, embonic acid, 4,4'-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7rrq8qz38n, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 7rrq8qz38n, pamoic acid 98+% hplc, 4,4'-methylen-bis-3-hydroxy-2-naphthoesaeure | |
CHEBI:50186 | |
OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1 |
Specifica
130-85-8 | |
99% | |
MFCD00004079 | |
14,7005 | |
Soluble in nitrobenzene, pyridine | |
OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1 | |
388.38 | |
CHEBI:50186 | |
99% |
>300°C | |
C23H16O6 | |
901319 | |
pamoic acid, embonic acid, 4,4'-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7rrq8qz38n, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 7rrq8qz38n, pamoic acid 98+% hplc, 4,4'-methylen-bis-3-hydroxy-2-naphthoesaeure | |
WLJNZVDCPSBLRP-UHFFFAOYSA-N | |
4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid | |
8546 | |
388.38 | |
Pamoic Acid |
Sicurezza e movimentazione
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 204-998-0
missing translation for 'rtecsNumber' : QL2180000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.