missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Panoramasque™ Full-Face Air Purifying Respirator (APR)
The Panoramasque™ Full-Face APR is a Mono filter with Rd40 thread connection system that offers respiratory protection, depending on the canister type assembled on it.
194.00 € - 421.00 €
Specifications
Certifications (EN) | EN 136:1989 |
---|---|
Connection Type | 1/7 in. Rd40 Thread System |
Filter Type | Mono-Filter |
Product Type | Full-Face Air Purifying Respirator (APR) |
For Use With (Application) | Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry |
Product Code | Brand | Facepiece Material | Material (Lens) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Facepiece Material | Material (Lens) | Includes | Price | Quantity & Availability | |||
12612923
|
Honeywell™
1710395 |
EPDM | PMMA | With Speech Diaphragm (Metal) |
194.00 €
Each |
In Stock
Log in to see stock. |
|||
12652923
|
Honeywell™
1710987 |
Silicone | PMMA | With Speech Diaphragm (Metal) |
405.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
12662923
|
Honeywell™
1711000 |
EPDM | Specific for Chlorine Hazards | Without Speech Diaphragm | N/A | ||||
12672923
|
Honeywell™
1711001 |
EPDM | Laminated Glass for Chemical Hazards | Without Speech Diaphragm |
421.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
15952912
|
Honeywell™
1710980 |
Silicone | PMMA | Without Speech Diaphragm |
260.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
12602923
|
Honeywell™
1710394 |
EPDM | PMMA | Without Speech Diaphragm |
278.00 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
Description
- Product Type: Full-Face Air Purifying Respirator (APR)
- Material (Body): EPDM or Silicone
- Material (Lens): PMMA, Glass, Chlorine Hazard Specific
- Color: Black, Yellow
- Cartridge: Single Filter via Rd40 (1/7 in.) Connection
- Use with: EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters
- PPE Class: Variable (Dependant on Cartridge)
- Hazard Types: Particulates, Gas, Vapor, Smoke
- Applications: Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry
Specifications
EN 136:1989 | |
Mono-Filter | |
Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry | |
Yes |
1/7 in. Rd40 Thread System | |
Full-Face Air Purifying Respirator (APR) | |
EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title