Learn More
3M™ Paper Sheet 618, P240
3M™ Paper Sheet 618 is a silicon carbide mineral abrasive on a flexible backing with a Free-Cut anti-loading coating.
Brand:  3M™ 7000034040
Product Code. 15664036
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Provides a sharp, aggressive cut with a fine finish due to the silicon carbide abrasive mineral
  • Features a Free-Cut anti-loading coating
  • Contains a flexible backing
  • Hand Blocks and Randon Orbital Sanders
  • Size: 230 x 280 mm
Specifications
Specifications
Paper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Paper Sheet 618, P240 > Color: Light Gray