Learn More
Paper Titan Yellow EU Pharmacopoeia, Fisher Chemical™
Product Code. 12990554
Click to view available options
Quantity:
50 mL
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 50 mL
Analyte or Component Names Paper Titan Yellow