missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde, 96%, Thermo Scientific Chemicals

Product Code. 10351641
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
500 g
1 kg
3 kg
10 kg
Packaging:
Plastic Bucket
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Decomposition Information 260°C
Melting Point 120.0°C to 170.0°C
Color White
Density 1.03 (Air = 1.0)
pH 4.0 to 5.5 (10% suspension)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Linear Formula (CH2O)n
Quantity 25 g
Solubility Information Solubility in water: sparingly soluble. Other solubilities: the higher polymers are insoluble in water, insoluble in alcohol and ether
Vapor Pressure 1.2mmHg at 25°C
Percent Purity 96%
Grade Extra Pure
Physical Form Beads or Powder
Chemical Name or Material Paraformaldehyde, Extra Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Paraformaldehyde, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.