Learn More
Thermo Scientific Chemicals Paraformaldehyde fixative solution
84.92 € - 154.77 €
Specifications
Chemical Name or Material | Paraformaldehyde fixative solution |
---|---|
Physical Form | Liquid |
MDL Number | MFCD00133991 |
Solubility Information | Miscible with water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15434459
|
Thermo Scientific Chemicals
J61984.AK |
250 mL |
84.92 €
250mL |
In Stock
Log in to see stock. |
|||||
15444459
|
Thermo Scientific Chemicals
J61984.AP |
500 mL |
154.77 €
500mL |
In Stock
Log in to see stock. |
|||||
Description
Paraformaldehyde fixative solution finds application in proteins, peptides and the enzymes of low molecular weight.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Paraformaldehyde fixative solution | |
Liquid | |
MFCD00133991 | |
Miscible with water. |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
H317-H318-H351
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- Paraformaldehyde fixative solution
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Paraformaldehyde
RUO – Research Use Only