Learn More
R&D Systems™ Parameter Immunoassay Control Set 931 for Thromboxane B2
For use with catalog number KGE011
Brand:  R&D Systems™ QC179
Product Code. 30001915
  • 196.00 € / Each
Explore more special offers
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Control
Multi-species
Thromboxane B2
1 Each