missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Lab Thermometer

General purpose laboratory thermometer without calibration certificate.

Brand:  Brannan™ 44/002/7

 View more versions of this product

Product Code. 12736998

  • 4.60 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Filled with a brightly colored, mercury-free spirit. Brannan™ Partial Immersion Spirit Filled Thermometers use specially selected filling materials covering temperatures down to -200°C and feature a non-roll cap with suspension ring.

  • Lab thermometer
  • Safe non-mercury filling
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications

Specifications

Red Spirit
Organic filling
Glass
Red Spirit
230°F
0°F to 230°F
Equivalent to SAMA No. FP15
2°C
1.2 cm
Partial Immersion
305 mm
SAMA FP20
Laboratory Thermometer
0°F
0°F to 230°F
±2°C
76 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.