Learn More
3M™ Particulate Respirators
To help improve fit to wearer’s individual face shape and size. 3M™ Particulate Respirators are reusable and can be used for more than one shift.
59.00 € - 67.00 €
Specifications
Product Type | Particulate Respirator |
---|---|
For Use With (Application) | Metalworking, Chemical Processing, Construction, Pharmaceuticals, Ship Building, Chemical Processing, Construction, Pharmaceuticals |
Latex Free | Yes |
Shape | Cup |
Description
- High performance filter material ensures easy breathing through
- Flexible material helps form comfortably onto a wide range of face shapes and sizes helping to give a customized fit
- Innovative contoured design engineered to help provide a more secure seal around the nose
- Adjustable colour coded braided headbands with 4-point adjustment helps to provide a more secure feel
- Easy to clean for wearer hygiene using the 3M™ Face Seal Cleaning Wipe 105 (as per EN 149+A1)
- Collapse resistant: works well in hot humid environments
- EN 149 ‘R’ Rated. Reusable
- Cool Flow™ Valve provides effective exit of heat build up provides a cooler and more comfortable wear and helps to minimize the risk of fogging eyewear
Specifications
Particulate Respirator | |
Metalworking, Chemical Processing, Construction, Pharmaceuticals, Ship Building, Chemical Processing, Construction, Pharmaceuticals | |
Yes | |
Cup |