Learn More
Hecht Karl™ Pasteur Pipets
Manufactured from glass for liquids susceptible to leaching from plastic pipets. Karl Hecht™ Pasteur Pipets are ideal for transferring small quantities of liquid.
Brand:  Hecht Karl™ 40567002
 View more versions of this product
Product Code. 10717152
  • 21.00 € / Pack of 250
Explore more special offers
Description
Description
  • Uniform shape
  • Disposable
  • Constriction in upper section for cotton plug 
Specifications
Specifications
Tip Opening: 1 mm
250 Pack
Long Form
7 mm
230 mm