missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Pasteur pipettes made from Borosilikat glass 5.1
Pasteur pipettes are intended for single use in microbiological and serological laboratories
Brand: DWK Life Sciences 233301504
This item is not returnable.
View return policy
Description
Thanks to their material properties, Pasteur pipettes made of borosilicate glass 5.1 are a high-quality alternative to plastic pipettes and pipettes made of soda-lime glass.
- Borosilicate glass 5.1 is more stable than soda-lime glass and is therefore insensitive to rapid temperature changes and chemical stress.
- Material properties ensure chemical resistance to acid and alkaline solutions
- Nearly inert borosilicate glass minimizes interactions with the material and reduces the risk of sample contamination
- Pasteur pipettes with short or long extended, fine tip (150 and 230 mm)
- The filling volume is approx. 2 ml.
- Natural rubber teat can be ordered separately.
Specifications
Not Autoclavable | |
Borosilicate Glass (5.1) | |
4 x 250 |
6.95 mm | |
Single use in microbiological and serological laboratory applications; Pipetting liquids; Dropwise dosing of liquids and reagents; Distribution of heated liquid quantities | |
150 mm |