missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Patent Blue VF, pure, indicator grade

Product Code. 15274664
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-17-9
Molecular Formula C27H31N2NaO6S2
Molecular Weight (g/mol) 566.66
MDL Number MFCD00012119
InChI Key SJEYSFABYSGQBG-UHFFFAOYSA-M
Synonym Acid Blue 1, C.I. 42045, αzurine 2G, Sulphan blue
PubChem CID 8507
ChEBI CHEBI:34906
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]benzene-1,3-disulfonate
SMILES [Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: 40g/L (95°C), 30 mg/l in egme, 7 mg/l in etoh
Infrared Spectrum Authentic
Formula Weight 566.67
Grade Pure
Suitability suitable as acid-base indicator
UV Spectrometry Complies
Color Blue/Green to Purple
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Patent Blue VF

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'