Learn More
Thermo Scientific™ PathoDx Strep Grouping Universal Kit
Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit.
187.00 €
Specifications
Control Sets | Positive |
---|---|
Detectable Analytes | beta-hemolytic streptococci (Groups A, B, C, F, G) |
Type | Streptococcus Test Kit |
Includes | Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
12791770
|
Thermo Scientific™
R62076 |
PathoDX™ Strep Universal Grouping Kit | 60 Tests/Kit |
187.00 €
60 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit. This kit is a set of components to be used with selected, separately purchased PathoDxStrep Grouping Reagents designed to identify beta-haemolytic streptococci of Lancefield Groups A, B, C, F or G from primary culture plates.This accessory kit contains: Extraction reagent 1; Extraction reagent 2; Extraction reagent 3; Positive control; Reaction cards; Stirrers.
The PathoDx procedure utilizes a latex agglutination method in conjunction with a nitrous acid extraction procedure.1,2 The IgG coupled to the latex is highly specific for a given streptococcal group antigen. This method offers significant advantages over other streptococcal grouping procedures in terms of rapidity, simplicity, and convenience.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Positive | |
beta-hemolytic streptococci (Groups A, B, C, F, G) | |
Streptococcus Test Kit | |
Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides |
Safety and Handling
- PathoDX STREP Grouping Universal Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 1 A
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium nitrite, Acetic acid, Carbonic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.