missing translation for 'onlineSavingsMsg'
Learn More

Intelsius™ PathoPak

PathoPak sample transport packaging is certified for Category A and B shipments, meeting IATA, ADR, and CFR 49 (DOT) transport regulations, providing protection for your samples regardless of their specimen classification.

Brand:  Intelsius™ PP011

Encompass_Preferred

 View more versions of this product

Product Code. 15645609

  • POA / Pack of 20

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A Strong, durable and highly protective shipping solution for UN3373 category B biological samples.
  • Suitable for shipping a wide range of primaries including:
    • Blood Tubes
    • Specimen Containers
    • Swabs
  • Components Supplied:
    • 95kPa IATA compliant secondary rigid pressure vessel
    • Outer carton printed in compliance with transport regulations
    • Absorbent
    • Bubble wrap
    • Infectious class label
    • Security Seal label
Specifications

Specifications

UN3373
Sample Transport Packaging
90 x 170 mm (3.5 x 6.7 in.)
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.