missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intelsius™ PathoSorb Absorbent Pouch
Protect and separate sample vials with this PathoSorb Absorbent sample Pouch.

POA
Specifications
Certifications/Compliance | UN3373, UN2900, UN2814 |
---|---|
Type | Absorbent Sample Pouch |
Product Code | Brand | Capacity | Dimensions (L x W) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Dimensions (L x W) | Includes | Price | Quantity & Availability | |||
17308589
![]() |
Intelsius™
AB005 |
152 mL | 225 x 125 mm (8.9 x 4.9 in.) | 4 Bays |
POA
Pack of 200 |
N/A | |||
17328589
![]() |
Intelsius™
AB008 |
215 mL | 325 x 125 mm (12.8 x 4.9 in.) | 6 Bays |
POA
Pack of 200 |
N/A | |||
Description
- Keeps sample tubes safe and secure
- Lint free
- Suitable in supporting UN3373 Category B and UN2900/UN2814 Category A transport regulations
Specifications
UN3373, UN2900, UN2814 | |
Absorbent Sample Pouch |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Intelsius™ PathoSorb Absorbent Pouch