missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD81 Isolation Kit for FABian™
IBA Lifesciences™ CD81 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
Brand: IBA Lifesciences 6-6015-010
This item is not returnable.
View return policy
Description
- The kit is used for the selection of PBMCs from whole blood or buffy coat in conjunction with the FABian™ system.
- It includes CD81 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
- Each kit is sufficient for one FABian™ run to isolate cells from 9mL whole blood.
Specifications
Isolation of label-free cells | |
2°C to 8°C | |
9 mL Whole Blood |
Cell Isolation Kit | |
Human Whole Blood | |
10 Kits, Human |