Learn More
Gibco™ PBS, pH 7.4

PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 10010023
33.59 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. We offer a variety of PBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This PBS is manufactured as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature

Specifications
No Sodium Pyruvate | |
Clear | |
1 X | |
Phosphate Buffered Saline (PBS) | |
Phosphate Buffered Saline | |
280 - 315 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Animal Origin-Free | |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
|
Gibco™ | |
500 mL | |
Washing Cells Before Dissociation; Diluting Cells for Counting and Preparing Reagents; Transporting Cells or Tissue | |
7.4 | |
Room Temperature | |
Liquid |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2572130 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2494223 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2494226 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2562446 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2436334 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.