missing translation for 'onlineSavingsMsg'
Learn More

PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™

Product Code. p-27643
Click to view available options
Quantity:
500mL
1L
4L
20L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly Cube
Unit Size:
1 L
20 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 10649743

Brand: Fisher BioReagents BP39920

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Commonly used buffer

  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.119M phosphates
  • pH of a 1X solution made from this concentrated solution is ∼7.4. To prepare a 1X solution, mix one volume of Fisher BioReagents 10X PBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Concentration 10 X
Physical Form Liquid
Quantity 20L
Solution Type Phosphate Buffered Saline
Packaging Poly Cube
Grade Biochemical
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.