missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS Tablets

Phosphate-buffered saline (PBS) is a balanced salt formulation used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Brand:  Gibco™ 18912014

Product Code. 11510546

  • 178.00 € / 100 tablets

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Phosphate-buffered saline (PBS) is a balanced salt formulation used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Each 5 g Gibco™ PBS Tablet should be dissolved in 500 mL of distilled water. The pH will be 7.45 and requires no adjustment.

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.

These PBS Tablets are manufactured as follows:
Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Phenol Red, No Magnesium, No Phenol Red
1 X
Phosphate Buffered Saline (PBS)
7.45
Solid
Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Gibco™
100 tablets
Room Temperature
cGMP-compliant under the ISO 13485 standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ PBS Tablets > 100 tablets

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.