Learn More
PCDHB12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14020-1-AP-20UL
 View more versions of this product
Product Code. 16832484
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.
Specifications
Specifications
PCDHB12
Polyclonal
Unconjugated
PCDHB12
PCDH beta 12, PCDH BETA12, PCDHB12, protocadherin beta 12
Rabbit
Antigen Affinity Chromatography
RUO
56124, 93883
-20°C
Liquid
Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y5F1
PCDHB12
PCDHB12 Fusion Protein Ag5135
20 μL
Primary
Mouse, Human