missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ 8-Strip PCR Caps, BIO-CERT™ PCR QUALITY

Product Code. p-6814344 Shop All Brand Products
Click to view available options
Color:
Blue
Clear
Green
Rose
Yellow
For Use With (Equipment):
Microplates
Thermal cycler
qPCR plates
Unit Size:
Pack of 125
Pack of 300
Format:
Domed
Flat
This item is not returnable. View return policy

Product Code. 10608131

Brand: BRAND™ 781340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BRAND™ PCR cap strips ensure reliable closures for PCR strips and plates, safeguarding your samples against evaporation and contamination.

  • Exceptional Quality and Purity: BIO-CERT™ PCR QUALITY ensures caps are free from human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
  • Secure Sealing: Designed to reliably seal PCR strips and plates with 24-, 48-, or 96-wells, effectively protecting your samples from evaporation and contamination.
  • Perfect Compatibility: Optimally shaped to match BRAND™ PCR tube strips and plates, ensuring easy opening and closing. They are also compatible with most other manufacturers, offering flexibility for your workflow.
  • Contamination Prevention: Equipped with side tabs for precise positioning and a small opening attachment on each cap to prevent direct contact with the interior.
  • qPCR-Ready Transparency: Highly transparent flat caps enable accurate fluorescence measurement, making them ideal for qPCR applications.
  • Enhanced Sealing with Domed Caps: Domed caps provide a stronger sealing effect when used with thermal cyclers lacking pressure sensors.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Equipment) Thermal cycler
Shape Domed
Sterility Non-sterile
Material Polypropylene
Dnase Free Yes
RNase Free Yes
Closure Type 8-strip Domed Cap
Autoclavable Autoclavable
Closure Included Yes
Disposable Single-use
Product Type Cap Strip
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.