missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ PCR-Cooler
Innovative solution for protecting, transporting and storing sensitive samples as well as for safely stopping reactions.
Brand: Eppendorf™ 3881000015
117.80 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
- Maintains a sample temperature of 0°C for over an hour and then changes color when the temperature has exceeded 7°C
- Superior to an ice bath, it is easy to handle, space saving and free of contamination, which makes it the perfect preparation for PCR reactions
- Temperature of single tubes is indicated by the color of the individual bores
- Samples are prevented from freezing by the specially developed cone contour
- Cooling aggregate in the universal 96 well format for 0.2mL and 0.5mL PCR tubes, for tube strips, plates and individual tubes
Specifications
PCR Reaction Setup | |
Blue, Pink | |
96 |
PCR Cooler | |
0.2 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction