missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PCR™ Cooler, 96-Well, 2 Pack
The PCR™ Cooler is ideal for thermosensitive applications, PCR, or cell-based assays, as well as gentle sample thawing and stopping of reactions. The unit also provides a dependable cooling station for pipetting and sample storage. Perfect for transporting samples and reactions between preparation, thermal cycling and post-PCR procedure areas.
Brand: Fisherbrand™ 120727F
96.17 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
- Modern patented design cools faster in freezers and stays cold longer on the bench top
- PCR™ Cooler maintains a sample temperature of 0 °C (with 2 hours pre-cooling at -20 °C) for over an hour
- When the temperature ceiling of 6 °C has been exceeded, users are signaled by a contrasting color change of the cooler, or even a single well
- Cools industry standard format 0.2 mL and 0.5 mL PCR™ tubes, strips and single format, as well as most 96 well plates, in both standard and low profile well shape
- Better alternative then ice baths as dry incubation reduces the risk of contamination
- Freeze cooler in an upright position enabling cold storage of tubes
- Well design prohibits the freezing of samples
- Fully sealed and sonic welded cooler keeps coolant contained offering worry free usage
- Save space with compact footprint and stacking ability
- The lid is ambidextrous; it can be switched to accommodate left or right-hand orientation and is detachable
- Robust cooler made from engineered polypropylene and the lid is made from polycarbonate
- US Patent Pending, Community Design
- No: 004101756-0001-0002
- Two-Year Warranty
Specifications
±1°C | |
Lid, Rack | |
Tubes | |
1.9 cm | |
4.8 cm | |
Fisherbrand™ PCR™ Cooler, 96-Well, 2 Pack | |
Purple, Pink | |
.27 kg (0.6 lb.) |
0°C to 5°C | |
Shakers, Mixers, Vortexers | |
Purple and Pink | |
14.8 cm | |
PCR Applications | |
2 | |
PCR Rack |