missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PCR Master Mix (2X)

Thermo Scientific PCR Master Mix is a 2X concentrated solution of Taq DNA Polymerase, dNTPs, and all of the components required for PCR, except DNA template and primers.

128.00 € - 495.00 €

Specifications

GC-Rich PCR Performance Low
Polymerase Taq DNA Polymerase
Reaction Speed Standard
Product Type PCR Master Mix
For Use With (Application) Standard PCR
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
11853913
View Documents
Thermo Scientific™
K0171
200 Reactions
128.00 €
200 reactions
Please to purchase this item. Need a web account? Register with us today!
11863913
View Documents
Thermo Scientific™
K0172
1000 Reactions
495.00 €
1000 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific PCR Master Mix is a 2X concentrated solution of Taq DNA Polymerase, dNTPs, and all of the components required for PCR, except DNA template and primers. This pre-mixed formulation saves time and reduces contamination due to a reduced number of pipetting steps required for PCR set up. The mix is optimized for efficient and reproducible PCR.

Highlights

  • Convenient, ready-to-use mix
  • Thermostable—half life is more than 40 min. at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • High throughput PCR
  • DNA labeling

Note

The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle, as determined by a modified method described in. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.

Specifications

Specifications

Low
Standard
Standard PCR
1X
3'-A
DNA
Taq DNA Polymerase
PCR Master Mix
2X
No
SuperMix or Master Mix
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.