Learn More
Tocris Bioscience™ PD 90780
Inhibits NGF binding to p75NTR
Brand:  Tocris Bioscience™ 5101/10
222.89 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18737088
  • 252.00 € / 10mg
Explore more special offers
Description
Description
PD 90780 is a a nonpeptide inhibitor of NGF binding to p75NTR (IC50 ∽ 1 μM in CHO cells); binds NGF, not p75NTR. Affinity maybe altered by the presence of TrkA receptors. Neuroprotective.
Specifications
Specifications
>98%
PD 90780
PD 90780
10 mg
362.34