missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PD MiniTrap™ G-25 Sample Preparation Columns
Use for cleanup of biomolecules with a molecular weight >5000 Mr in sample volumes up to 0.5mL. Cytiva Life Sciences™ D MiniTrap™ G-25 Sample Preparation Columns are convenient and prepacked.
Brand: Cytiva 28-9180-07
Includes: Four centrifuge adapters
Description
- Developed for a wide range of applications: desalting, buffer exchange, removal of low-molecular-weight compounds, etc.
- Equally suited for gravity flow or centrifugation protocols
- Accommodates sample volumes of 0.1 to 0.5mL for gravity, 0.2 to 0.5mL with centrifugation
- High recoveries of 70 to 95%, depending on biomolecule
- High desalting capacity of >90%
Specifications
PD MiniTrap Sephadex G-25 Gravity Column | |
0.38 in. | |
1.2 in. | |
>100cm/hr. | |
Polyethylene | |
50μm in dry form, 38 to 235μm in wet form | |
4° to 30°C | |
Buffer exchange and cleanup of biological samples, such as proteins and carbohydrates |
2.1mL | |
9.7mm | |
28mm | |
Four centrifuge adapters, instructions | |
2 to 13 | |
Cross-linked Dextran | |
Gravity Column |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva PD MiniTrap™ G-25 Sample Preparation Columns > Sample volume 0.2 to 0.5mL
Spot an opportunity for improvement?Share a Content Correction