missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE6H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18151-1-AP-150UL
This item is not returnable.
View return policy
Description
Participates in processes of transmission and amplification of the visual signal. cGMP-PDEs are the effector molecules in G-protein-mediated phototransduction in vertebrate rods and cones.Specifications
PDE6H | |
Polyclonal | |
Unconjugated | |
PDE6H | |
GMP PDE gamma, PDE6H, RCD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114248, 5149, 78600 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61249, Q13956 | |
PDE6H | |
PDE6H Fusion Protein Ag12847 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |