missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13404-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the dephosphorylation and concomitant reactivation of the alpha subunit of the E1 component of the pyruvate dehydrogenase complex.Specifications
PDP2 | |
Polyclonal | |
Unconjugated | |
PDP2 | |
KIAA1348, PDP 2, PDP2, PDPC 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246311, 382051, 57546 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88484, Q9P2J9 | |
Pdp2 | |
PDP2 Fusion Protein Ag4257 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction