Learn More
asecos™ PE IBC Station with Filling Area and Galvanised Grid
The PE IBC Station is approved for the safe storage of water pollutants.
Brand: asecos™ 24156
This item is not returnable.
View return policy
Description
- Highly resistant against oil, acid, alkali, etc.
- Easy and safe in-house transport of the empty sump by forklift truck
Specifications
Blue | |
1000 L | |
IBC Spill Pallet | |
1280.00 mm | |
1280.00 mm | |
860.00 mm | |
1910.00 mm | |
115 kg |
264.2 gal. | |
Polyethylene | |
50.39 in. | |
50.39 in. | |
33.86 in. | |
75.20 in. | |
253.53 lb. |