missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ LDPE Grip Stoppers
Manufactured from Low-density polyethylene (LDPE) BRAND™ LDPE Grip Stoppers are used with sample and sedimentation tubes. The dual-ringed stopper prevents sample leakage. Ridged edge provides easy gripping and handling.
Brand: BRAND™ 114730
30.07 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from Low-density polyethylene (LDPE)
- For use with sample and sedimentation tubes.
- Dual-ringed stopper prevents sample leakage.
- Ridged edge provides easy gripping and handling.
Specifications
12mm | |
BRAND™ Sample Tubes 114750, 114760 | |
11.5 to 12 Dmm | |
Grip Stopper |
Sample and sedimentation tubes | |
1000/Ea. | |
LDPE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction