Learn More
Fisherbrand™ PE Plug for Shell Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 12 08 0546
This item is not returnable.
View return policy
Description
- PE Plug
- Transparent
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Transparent | |
Polyethylene | |
Plug |
12 mm | |
100 | |
ND12 |