missing translation for 'onlineSavingsMsg'
Learn More

asecos™ PE Storage Can with Dosing Tap

The PE Storage Can is ideal for the safe storage and internal transport of liquids, such as acids or lyes.

Brand:  asecos™ 40095

 View more versions of this product

Product Code. 17806336

  • 85.00 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With fine dosing tap made of stainless steel (1.4301), incl. vent, PTFE seal
  • Incl. multilingual safety sticker to designate the contents
Specifications

Specifications

0.3 gal.
Yellow
3.94 in.
9.06 in.
1.10 lb.
1 L
Polyethylene
100.00 mm
230.00 mm
0.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers