Learn More
Fisherbrand™ PE Tank with Level Sensor
Holds 30 or 60L. Fisher Scientific™ PE Tank with Level Sensor is equipped with a CO2 scavenger to capture and remove carbon dioxide.
Brand:  Fisherbrand™ RATANK030
 View more versions of this product
Product Code. 15479679
  • 984.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Level sensor indicates amount of liquid in tank
  • Tank vent filter includes CO2 scavenger to protect purified water from atmospheric degradation.
  • Available in 30L or 60L
Specifications
Specifications
30 L
Tank, PE
Polyethylene
Accu500
Water purification