Learn More
Lenz Laborglasinstrumente™ Pear-shaped Evaporating Flask with Conical Ground Joint
Especially for rotary evaporators, with center neck and conical joint, pear-shaped, with inscription label.
Brand:  Lenz Laborglasinstrumente™ 03041413
 View more versions of this product
Product Code. 11973381
  • 187.00 € / Pack of 10
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Duran Glass
Rotary Evaporators
Pear
33 mm
Evaporating Flask
14/23
10 mL