missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEBP1 Mouse anti-Human, Mouse, Rat, Clone: 2D4D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66438-1-IG-150UL
This item is not returnable.
View return policy
Description
PBP binds ATP, opioids and phosphatidylethanolamine, exhibiting a lower affinity for phosphatidylinositol and phosphatidylcholine. This serine protease inhibitor inhibits thrombin, neuropsin and chymotrypsin but not trypsin, tissue type plasminogen activator and elastase. PBP contains hippocampal cholinergic neurostimulating peptide (HCNP), which may be involved in the function of the presynaptic cholinergic neurons of the central nervous system. HCNP increases the production of choline acetyltransferase but not acetylcholinesterase.Specifications
PEBP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30086, P31044, P70296 | |
PEBP1 | |
PEBP1 Fusion Protein Ag0864 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2D4D11 | |
Unconjugated | |
PEBP1 | |
HCNP, HCNPpp, Neuropolypeptide h3, PBP, PEBP, PEBP 1, PEBP1, Prostatic binding protein, Raf kinase inhibitor protein, RKIP | |
Mouse | |
Protein G | |
RUO | |
23980, 29542, 5037 | |
-20°C | |
Liquid |