missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Pedal Container with Door and Lid, 90 L

90 L Pedal Container with Door and Lid allows easy bag removal thanks to side opening and is easy to move around.

Brand:  Gilac™ G614823

 View more versions of this product

Product Code. 17978741

  • 425.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Polaris pedal container.
  • Equipped with two antimark wheels (diameter 120 mm).
  • Easy to move around.
  • Easy bag removal thanks to side opening.
  • Hands-free operation for sanitary disposal.
  • Easy to clean.
  • Colored lids for selective waste sorting.
Specifications

Specifications

90 L
Green
Polypropylene
23.8 gal.
Not Autoclavable
Pedal Container with Lid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.