Learn More
Hughes Pedestal Mounted Closed Bowl Eye/Face Wash
Pedestal mounted emergency eye/face wash.
Brand:  Hughes HEWP0045GS
Product Code. 17722338
  • 1125.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pedestal mounted emergency eye/face wash
  • This ANSI compliant model has an integral lid to protect the bowl and diffusers against dust and debris
  • Designed to be quick and easy to operate with high flow rate aerated diffusers for improved washing
Specifications
Specifications
46 cm
Pedestal
32 cm
Stainless Steel
Emergency Shower