Learn More
Hughes Pedestal Mounted Open Bowl Eye/Face Wash
Open stainless steel bowl, pedestal-mounted emergency eye/face wash featuring twin spray nozzles protected by dust covers which lift off upon activation via the push plate.
Brand:  Hughes HEWP0085SS
Product Code. 17732338
  • 613.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Open stainless steel bowl, pedestal-mounted emergency eye/face wash featuring twin spray nozzles protected by dust covers which lift off upon activation via the push plate
  • The flow regulator valve guarantees an optimum water flow
  • The eye wash diffusers are fitted with fine mesh strainers and individual flow controllers designed to give a gentle flow of aerated water at 12 liters per minute as per the ANSI standard
  • This unit is intended for indoor use but can be located outdoors where there is no risk of overheating or freezing
Specifications
Specifications
35 cm
Pedestal
38 cm
Stainless Steel
Emergency Shower