missing translation for 'onlineSavingsMsg'
Learn More

Manutan Pedestrian internal quick-hang PVC curtain, 2400mm x 2500mm

Brand:  Manutan A351941

Encompass_Preferred

Product Code. 12844329

  • 485.28 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Quick hang rail supplied in 984mm sections, with predrilled holes, for fixing to a wall face or under ceiling. PVC strips create a see through flexible curtain. Keeps heat in and reduces cold air loss in chill/freezer rooms for greater temperature control. Provides protection against rain and wind. Restricts insect and bird access. Forms a good noise barrier. Partial - 50% overlap. Easier access for pedestrians.
  • Door height: 2400mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Manutan Pedestrian internal quick-hang PVC curtain, 2400mm x 2500mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.