missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Peek Analytical Guard Replacement Cartridges
Brand: Hamilton™ 79377
This item is not returnable.
View return policy
Specifications
5 Pack | |
8 mm | |
Replacement Cartridges | |
PRP-X400 | |
PSDVB/Sulfonic Acid |
7 μm | |
5000 psi | |
3 mm | |
PEEK | |
Replacement Guard Cartridge |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hamilton™ Peek Analytical Guard Replacement Cartridges > Packing Material: PSDVB/Sulfonic Acid
Spot an opportunity for improvement?Share a Content Correction