Learn More
Cytiva PEEK Connector Luer/M6
Set of five (5) polyetheretherketone Luer Lock Male/M6 Female connectors for use with the ÄKTAcrossflow tangential flow filtration system.
Brand:  Cytiva 11-0026-43
Product Code. 11380142
  • 284.83 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
ÄKTAcrossflow is a fully automated tangential flow filtration system for process development and is suitable for filter screening and process optimization at small-scale as well as small-scale processing.
  • Polyetheretherketone (PEEK) connstruction
  • Connections: Luer male-M6 female
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Connector Luer
Set of 5
Black
Luer Male to M6 Female
PEEK
ÄKTAcrossflow Tangential Flow Filtration System