Learn More
Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/8 in. OD Tubing, 1/4-28 Flat-Bottom
Eliminate the need to flange tubing with this Standard Knurl Flangeless Male Nut.
Brand:  Idex 02007-66
Product Code. 11795908
  • 75.75 € / Pack of 10
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Removable and reusable—saving you money.
  • Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port.
  • Unlike the time required to flange tubing, it only takes a few seconds to change the components.
  • Select from various ferrule materials to suit your application.
Specifications
Specifications
Nut
10 Pk.
PEEK
Natural
For 1/8 in. 1/4 to 28 Thread Size