Learn More
Cytiva PEEK Male Connector
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
Brand:  Cytiva 18-1170-59
 View more versions of this product
Product Code. 11300072
  • 481.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Chromatography accessory
  • Complete Packsize 2
  • Select from a variety of connections sizes and internal diamters 
  • For use with chromatography tubing
  • Connectors made of Polyetheretherketone (PEEK)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Male Connector
3 x 5/16 in. Female
PEEK