Learn More
Vici Valco Instruments™ One-Piece Fingertight Column Coupler
Choose from a variety of coupler IDs, indicated by the color of the sleeve
Brand:  Vici Valco Instruments™ JR-26502
 View more versions of this product
Product Code. 12915011
  • 159.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-adjusting - zero dead volume for all fitting types
  • Four bore sizes
  • Biocompatible
  • Adapts automatically to fit all pilot lengths
Specifications
Specifications
One Piece Fingertight Column Coupler
Yellow
0.17 mm
PEEK